siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 15 | 2385 | ||
| NM_001290736.1 | NP_001277665.1 | 15 | 2396 | |||
| NM_028789.3 | NP_083065.1 | 5 | 1024 | |||
| GenBank mRNA | AK004898.1 | - | 5 | 1025 | ||
| BC059910.1 | - | 15 | 2396 | |||
| BC089378.1 | - | 6 | 1138 | |||

Unkl
Genomic Map