siRNA Details
| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028792.3 | NP_083068.1 | 4 | 1746 | ||
| GenBank mRNA | AK004913.1 | - | 4 | 1695 | ||
| AK154799.1 | - | 4 | 1795 | |||
| AK177448.1 | - | 1 | 189 | |||
| AK179417.1 | - | 1 | 189 | |||
| AK180091.1 | - | 1 | 189 | |||
| AK195959.1 | - | 1 | 189 | |||
| AK199151.1 | - | 1 | 189 | |||
| AK199643.1 | - | 1 | 189 | |||
| AK216631.1 | - | 1 | 189 | |||
| AK216754.1 | - | 1 | 189 | |||
| AK216815.1 | - | 1 | 189 | |||
| BC006928.1 | - | 4 | 894 | 2655826 | ||
| BC086769.1 | - | 4 | 866 | 5707990 | ||

Josd1
Genomic Map