siRNA Details
| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004444.1 | - | 5 | 590 | ||
| AK076500.1 | - | 5 | 577 | |||
| AK151245.1 | - | 4 | 397 | |||
| AK158058.1 | - | 5 | 652 | |||
| BC033921.1 | - | 5 | 562 | 5068809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt9| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004444.1 | - | 5 | 590 | ||
| AK076500.1 | - | 5 | 577 | |||
| AK151245.1 | - | 4 | 397 | |||
| AK158058.1 | - | 5 | 652 | |||
| BC033921.1 | - | 5 | 562 | 5068809 | ||