siRNA Details
| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004444.1 | - | 6, 7 | 831 | ||
| AK076500.1 | - | 6, 7 | 818 | |||
| AK151245.1 | - | 5, 6 | 637 | |||
| AK158058.1 | - | 6, 7 | 893 | |||
| AK179782.1 | - | 1, 2 | 132 | |||
| AK219306.1 | - | 1, 2 | 121 | |||
| BC033921.1 | - | 6, 7 | 803 | 5068809 | ||

Nudt9
Genomic Map