siRNA Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145827.1 | NP_001139299.1 | 7, 8 | 1119 | ||
| NM_028800.3 | NP_083076.3 | 5, 6 | 765 | |||
| GenBank mRNA | AB102946.1 | - | 5, 6 | 596 | ||
| AK050808.1 | - | 1, 2 | 68 | |||
| AK154780.1 | - | 7, 8 | 1110 | |||
| AY336057.1 | - | 5, 6 | 750 | |||
| BC046981.1 | - | 5, 6 | 724 | 6478265 | ||

Stk40
Genomic Map