siRNA Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145827.1 | NP_001139299.1 | 13 | 2048 | ||
| NM_028800.3 | NP_083076.3 | 11 | 1694 | |||
| GenBank mRNA | AB102946.1 | - | 11 | 1525 | ||
| AK009154.1 | - | 6 | 912 | |||
| AK050808.1 | - | 7 | 997 | |||
| AK154780.1 | - | 13 | 2039 | |||
| AY336057.1 | - | 11 | 1679 | |||
| BC046981.1 | - | 11 | 1653 | 6478265 | ||

Stk40
Genomic Map