siRNA Details
| Gene Symbol: 2810403A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028814.3 | NP_083090.2 | 10 | 1094 | ||
| GenBank mRNA | AK012969.1 | - | 10 | 1071 | ||
| AK083018.1 | - | 10 | 1055 | |||
| AK170490.1 | - | 10 | 1089 | |||
| AK173057.1 | - | 10 | 1058 | |||
| BC006621.1 | - | 10 | 1069 | 3158104 | ||

2810403A07Rik
Genomic Map