siRNA Details
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028833.3 | NP_083109.2 | 6 | 601 | ||
| GenBank mRNA | AK006472.2 | - | 5 | 342 | ||
| AK029333.1 | - | 5 | 365 | |||
| BC059223.1 | - | 6 | 601 | 6417270 | ||
| BC065128.1 | - | 5 | 353 | |||

Iqce
Genomic Map