siRNA Details
| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004367.5 | NP_004358.2 | 3 | 438 | ||
| NM_031409.3 | NP_113597.2 | 3 | 701 | |||
| GenBank mRNA | AK223494.1 | - | 3 | 357 | ||
| AK293015.1 | - | 3 | 375 | |||
| AY242126.1 | - | 1 | 133 | |||
| BC037960.1 | - | 3 | 339 | 5748098 | ||
| DN996219.1 | - | 3 | 426 | |||
| U60000.1 | - | 1 | 133 | |||
| U68030.1 | - | 4 | 397 | |||

CCR6
Genomic Map