siRNA Details
| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028846.5 | NP_083122.1 | 6 | 504 | ||
| GenBank mRNA | AF449715.1 | - | 6 | 455 | ||
| AK029890.1 | - | 401 | ||||
| AK042612.1 | - | 6 | 421 | |||
| AK047242.1 | - | 6 | 425 | |||
| AK054279.1 | - | 6 | 447 | |||
| AK087412.1 | - | 6 | 415 | |||
| AK163663.1 | - | 6 | 414 | |||
| AK173083.1 | - | 6 | 406 | |||
| BC079674.1 | - | 393 | 30531994 | |||

Usp20
Genomic Map