siRNA Details
| Gene Symbol: Chic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006962.1 | - | 3 | 623 | ||
| AK172289.1 | - | 3 | 616 | |||
| BC048435.1 | - | 4 | 376 | |||
| BC116971.1 | - | 3 | 433 | 40126290 | ||
| BC116973.1 | - | 3 | 433 | 40126292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chic2| Gene Symbol: Chic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006962.1 | - | 3 | 623 | ||
| AK172289.1 | - | 3 | 616 | |||
| BC048435.1 | - | 4 | 376 | |||
| BC116971.1 | - | 3 | 433 | 40126290 | ||
| BC116973.1 | - | 3 | 433 | 40126292 | ||