siRNA Details
| Gene Symbol: Mettl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028865.3 | NP_083141.3 | 3 | 477 | ||
| GenBank mRNA | AK003450.1 | - | 3 | 323 | ||
| AK135294.1 | - | 438 | ||||
| AK160133.1 | - | 3 | 320 | |||
| BC026936.1 | - | 3 | 392 | 4021089 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK016897.1 | - | 438 | Chr. - - [-] on Build | ||

Mettl23
Genomic Map