siRNA Details
| Gene Symbol: Lrrtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028880.3 | NP_083156.2 | 2 | 2126 | ||
| GenBank mRNA | AK014545.1 | - | 2 | 2128 | ||
| AK053345.1 | - | 2 | 2116 | |||
| AK053364.1 | - | 2 | 1750 | |||
| AK080836.1 | - | 2 | 2157 | |||
| AK136857.1 | - | 1 | 1625 | |||
| BC027803.1 | - | 2 | 2008 | 5321979 | ||

Lrrtm1
Genomic Map