siRNA Details
| Gene Symbol: Tldc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028883.2 | NP_083159.1 | 8 | 1746 | ||
| GenBank mRNA | AK014579.1 | - | 8 | 1732 | ||
| AK079186.1 | - | 8 | 1746 | |||
| AK079239.1 | - | 8 | 1749 | |||
| AK173215.1 | - | 6 | 1048 | |||
| AK212803.1 | - | 1 | 246 | |||
| BC030874.1 | - | 7 | 1686 | 4224306 | ||

Tldc1
Genomic Map