siRNA Details
| Gene Symbol: Gcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028900.4 | NP_083176.3 | 1 | 1339 | ||
| GenBank mRNA | AK016531.1 | - | 1288 | |||
| AK032253.1 | - | 1 | 1339 | |||
| AK181400.1 | - | 1 | 250 | |||
| BC066807.1 | - | 1 | 1308 | 30101518 | ||

Gcc1
Genomic Map