siRNA Details
| Gene Symbol: 4932438H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163695.1 | NP_001157167.1 | 2 | 902 | ||
| NM_028905.3 | NP_083181.1 | 3 | 1150 | |||
| GenBank mRNA | AK016554.1 | - | 3 | 1151 | ||
| AK035767.1 | - | 2 | 892 | |||

4932438H23Rik
Genomic Map