siRNA Details
| Gene Symbol: Dpp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028906.2 | NP_083182.2 | 6 | 1020 | ||
| GenBank mRNA | AK016546.1 | - | 6 | 1059 | ||
| AK029788.1 | - | 5 | 2953 | |||
| AK155605.1 | - | 6 | 1064 | |||
| BC043124.1 | - | 6 | 1020 | 6410075 | ||
| BC059222.1 | - | 6 | 1020 | 6410075 | ||

Dpp8
Genomic Map