siRNA Details
| Gene Symbol: Dpp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028906.2 | NP_083182.2 | 21 | 3574 | ||
| GenBank mRNA | AK029788.1 | - | 19 | 5445 | ||
| AK171895.1 | - | 8 | 1644 | |||
| BC043124.1 | - | 21 | 3574 | 6410075 | ||
| BC059222.1 | - | 21 | 3574 | 6410075 | ||

Dpp8
Genomic Map