siRNA Details
| Gene Symbol: Sgms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028943.5 | NP_083219.2 | 2 | 732 | ||
| GenBank mRNA | AK016659.1 | - | 2 | 746 | ||
| BC117782.2 | - | 2 | 736 | 40092081 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgms2| Gene Symbol: Sgms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028943.5 | NP_083219.2 | 2 | 732 | ||
| GenBank mRNA | AK016659.1 | - | 2 | 746 | ||
| BC117782.2 | - | 2 | 736 | 40092081 | ||