siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290148.1 | NP_001277077.1 | 12 | 1145 | ||
| NM_028964.4 | NP_083240.2 | 19 | 2194 | |||
| GenBank mRNA | AF399755.1 | - | 12 | 1100 | ||
| AK017102.1 | - | 12 | 1146 | |||

Snx29
Genomic Map