siRNA Details
| Gene Symbol: 8430419L09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028982.4 | NP_083258.2 | 8 | 1329 | ||
| GenBank mRNA | AK018428.1 | - | 8 | 1273 | ||
| AK039401.1 | - | 8 | 1265 | |||
| BC079676.1 | - | 8 | 1256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430419L09Rik| Gene Symbol: 8430419L09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028982.4 | NP_083258.2 | 8 | 1329 | ||
| GenBank mRNA | AK018428.1 | - | 8 | 1273 | ||
| AK039401.1 | - | 8 | 1265 | |||
| BC079676.1 | - | 8 | 1256 | |||