siRNA Details
| Gene Symbol: Mgme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289630.1 | NP_001276559.1 | 2 | 578 | ||
| NM_001289631.1 | NP_001276560.1 | 2 | 249 | |||
| NM_028984.5 | NP_083260.1 | 2 | 307 | |||
| GenBank mRNA | AK018387.1 | - | 2 | 236 | ||
| AK031535.1 | - | 2 | 177 | |||
| AK043140.1 | - | 2 | 246 | |||
| AK049565.1 | - | 2 | 448 | |||
| AK159286.1 | - | 2 | 229 | |||
| AK167919.1 | - | 2 | 236 | |||
| AK187747.1 | - | 1 | 152 | |||
| AK188488.1 | - | 1 | 154 | |||
| BC063088.1 | - | 2 | 222 | 6329248 | ||
| BY136513.1 | - | 2 | 296 | |||

Mgme1
Genomic Map