siRNA Details
| Gene Symbol: Mgme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289630.1 | NP_001276559.1 | 2 | 655 | ||
| NM_001289631.1 | NP_001276560.1 | 2 | 326 | |||
| NM_028984.5 | NP_083260.1 | 2 | 384 | |||
| GenBank mRNA | AK018387.1 | - | 2 | 313 | ||
| AK031535.1 | - | 2 | 254 | |||
| AK043140.1 | - | 2 | 323 | |||
| AK049565.1 | - | 2 | 525 | |||
| AK159286.1 | - | 2 | 306 | |||
| AK167919.1 | - | 2 | 313 | |||
| AK187747.1 | - | 1 | 229 | |||
| BC063088.1 | - | 2 | 299 | 6329248 | ||

Mgme1
Genomic Map