siRNA Details
| Gene Symbol: Cd200r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027578.1 | NP_081854.1 | 5 | 771 | ||
| NM_029018.4 | NP_083294.2 | 6 | 990 | |||
| GenBank mRNA | AK014637.1 | - | 5 | 745 | ||
| AK040868.1 | - | 6 | 978 | |||
| AY230199.1 | - | 6 | 964 | |||
| BC106838.2 | - | 5 | 717 | 40044906 | ||

Cd200r3
Genomic Map