siRNA Details
| Gene Symbol: Tmem81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029025.3 | NP_083301.1 | 2 | 1386 | ||
| GenBank mRNA | AK015245.1 | - | 2 | 1387 | ||
| AK045585.1 | - | 2 | 1326 | |||
| BC120863.1 | - | 2 | 1146 | 40129786 | ||
| BC132045.1 | - | 2 | 1146 | 40130322 | ||

Tmem81
Genomic Map