siRNA Details
| Gene Symbol: Spdyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029048.3 | NP_083324.3 | 6, 7 | 929 | ||
| GenBank mRNA | AK015385.1 | - | 5, 6 | 739 | ||
| AK015441.1 | - | 6, 7 | 930 | |||
| BC141200.1 | - | 5, 6 | 739 | 9055812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spdyb| Gene Symbol: Spdyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029048.3 | NP_083324.3 | 6, 7 | 929 | ||
| GenBank mRNA | AK015385.1 | - | 5, 6 | 739 | ||
| AK015441.1 | - | 6, 7 | 930 | |||
| BC141200.1 | - | 5, 6 | 739 | 9055812 | ||