siRNA Details
| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127191.2 | NP_001120663.1 | 7 | 1446 | ||
| NM_029068.4 | NP_083344.3 | 8 | 1566 | |||
| GenBank mRNA | AK032055.1 | - | 8 | 1523 | ||
| BC070431.1 | - | 1886 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx16| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127191.2 | NP_001120663.1 | 7 | 1446 | ||
| NM_029068.4 | NP_083344.3 | 8 | 1566 | |||
| GenBank mRNA | AK032055.1 | - | 8 | 1523 | ||
| BC070431.1 | - | 1886 | ||||