siRNA Details
| Gene Symbol: C2cd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109688.2 | NP_001103158.1 | 7 | 979 | ||
| NM_001286578.1 | NP_001273507.1 | 6 | 807 | |||
| NM_029081.3 | NP_083357.2 | 7 | 979 | |||
| NM_029897.2 | NP_084173.1 | 7 | 979 | |||
| GenBank mRNA | AK029825.1 | - | 7 | 899 | ||
| AK143732.1 | - | 7 | 934 | |||
| AK162114.1 | - | 4 | 532 | |||
| AK163233.1 | - | 3 | 324 | |||
| AK172966.1 | - | 6 | 739 | |||
| BC027763.1 | - | 6 | 755 | 5354483 | ||
| BC053913.1 | - | 6 | 755 | 30053530 | ||
| CF916173.1 | - | 4 | 547 | |||

C2cd5
Genomic Map