siRNA Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029186.2 | NP_083462.2 | 5 | 977 | ||
| GenBank mRNA | AK015988.1 | - | 2 | 334 | ||
| AK031554.1 | - | 5 | 923 | |||
| AK052797.1 | - | 2 | 135 | |||
| AK151797.1 | - | 5 | 952 | |||
| AK161305.1 | - | 5 | 933 | |||
| AK170247.1 | - | 924 | ||||
| BC026385.1 | - | 3 | 1105 | |||
| BC031169.1 | - | 5 | 922 | |||
| BC049142.1 | - | 2 | 263 | |||
| BC058746.1 | - | 5 | 923 | 6399588 | ||

Tmem180
Genomic Map