siRNA Details
| Gene Symbol: 4930550C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029247.3 | NP_083523.1 | 3 | 199 | ||
| GenBank mRNA | AK016083.1 | - | 3 | 200 | ||
| BC115639.1 | - | 3 | 187 | 40102983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930550C14Rik| Gene Symbol: 4930550C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029247.3 | NP_083523.1 | 3 | 199 | ||
| GenBank mRNA | AK016083.1 | - | 3 | 200 | ||
| BC115639.1 | - | 3 | 187 | 40102983 | ||