siRNA Details
| Gene Symbol: 4930563D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029252.2 | NP_083528.1 | 1 | 1200 | ||
| GenBank mRNA | AK029653.1 | - | 1 | 1168 | ||
| BC147584.1 | - | 1 | 1181 | 9056574 | ||
| BC147622.1 | - | 1 | 1181 | 9056612 | ||

4930563D23Rik
Genomic Map