siRNA Details
| Gene Symbol: Slitrk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198865.1 | NP_942565.1 | 2 | 1020 | ||
| GenBank mRNA | AK173062.1 | - | 1 | 51 | ||
| BC065107.1 | - | 2 | 639 | 30357422 | ||
| BQ769394.1 | - | 1 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slitrk5| Gene Symbol: Slitrk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198865.1 | NP_942565.1 | 2 | 1020 | ||
| GenBank mRNA | AK173062.1 | - | 1 | 51 | ||
| BC065107.1 | - | 2 | 639 | 30357422 | ||
| BQ769394.1 | - | 1 | 413 | |||