siRNA Details
| Gene Symbol: Nop14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029278.2 | NP_083554.2 | 18 | 2610 | ||
| GenBank mRNA | AK148605.1 | - | 18 | 2568 | ||
| BC024998.1 | - | 18 | 2531 | 5012070 | ||
| BC043043.1 | - | 18 | 2553 | 5701430 | ||

Nop14
Genomic Map