siRNA Details
| Gene Symbol: Zfp821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | EU234026.1 | - | 676 | |||
| Gene Symbol: 1700008F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168369.1 | NP_001161841.1 | 15 | 1499 | ||
| NM_029292.3 | NP_083568.2 | 11 | 1009 | |||
| GenBank mRNA | AK005764.1 | - | 11 | 1010 | ||
| BC050962.1 | - | 14 | 1021 | |||
| BC061058.1 | - | 12 | 898 | |||

Zfp821
Genomic Map