siRNA Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290569.1 | NP_001277498.1 | 6 | 599 | ||
| NM_029377.2 | NP_083653.1 | 6 | 473 | |||
| GenBank mRNA | AK007858.1 | - | 6 | 474 | ||
| AY428767.1 | - | 6 | 323 | |||
| BC013546.1 | - | 6 | 455 | 4210417 | ||
| BY707863.1 | - | 6 | 473 | |||

Lin37
Genomic Map