siRNA Details
| Gene Symbol: Iqcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029408.2 | NP_083684.1 | 2 | 261 | ||
| GenBank mRNA | AK017032.1 | - | 2 | 261 | ||
| AK076996.1 | - | 2 | 268 | |||
| BC100441.1 | - | 2 | 250 | |||
| BC125429.1 | - | 2 | 255 | 40129944 | ||
| BC137644.1 | - | 2 | 255 | 8860663 | ||

Iqcd
Genomic Map