siRNA Details
| Gene Symbol: Zdhhc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029492.4 | NP_083768.4 | 8, 9 | 818 | ||
| GenBank mRNA | AK017158.1 | - | 8, 9 | 819 | ||
| AK031184.1 | - | 8, 9 | 777 | |||
| AK049270.1 | - | 8, 9 | 798 | |||
| AK140182.1 | - | 9, 10 | 815 | |||
| AK151840.1 | - | 8, 9 | 833 | |||
| AK163174.1 | - | 8, 9 | 816 | |||
| AK170017.1 | - | 8, 9 | 836 | |||
| AK170139.1 | - | 8, 9 | 759 | |||
| AY668951.1 | - | 9, 10 | 772 | |||
| BC019536.1 | - | 8, 9 | 835 | 4039734 | ||

Zdhhc20
Genomic Map