siRNA Details
| Gene Symbol: Zmym2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029498.3 | NP_083774.2 | 2 | 503 | ||
| GenBank mRNA | AK135499.1 | - | 3 | 616 | ||
| BC046507.1 | - | 3 | 549 | |||
| BC141403.1 | - | 2 | 479 | 9056016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmym2| Gene Symbol: Zmym2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029498.3 | NP_083774.2 | 2 | 503 | ||
| GenBank mRNA | AK135499.1 | - | 3 | 616 | ||
| BC046507.1 | - | 3 | 549 | |||
| BC141403.1 | - | 2 | 479 | 9056016 | ||