siRNA Details
| Gene Symbol: Gbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029509.4 | NP_083785.3 | 1675 | |||
| GenBank mRNA | BC031475.1 | - | 2591 | 2654170 | ||
| BC080665.1 | - | 6 | 1114 | |||
| DQ295175.1 | - | 9 | 1590 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbp8| Gene Symbol: Gbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029509.4 | NP_083785.3 | 1675 | |||
| GenBank mRNA | BC031475.1 | - | 2591 | 2654170 | ||
| BC080665.1 | - | 6 | 1114 | |||
| DQ295175.1 | - | 9 | 1590 | |||