siRNA Details
| Gene Symbol: Rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029519.3 | NP_083795.2 | 2 | 1112 | ||
| GenBank mRNA | AK075712.1 | - | 2 | 1072 | ||
| AK140318.1 | - | 2 | 1124 | |||
| BC043066.1 | - | 2 | 1083 | 5709337 | ||
| BC049084.1 | - | 2 | 1112 | 5706690 | ||
| BC053003.1 | - | 2 | 1112 | 5706690 | ||
| BC067194.1 | - | 2 | 1128 | |||

Rap2a
Genomic Map