siRNA Details
| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113327.2 | NP_001106798.1 | 4 | 521 | ||
| NM_001286584.1 | NP_001273513.1 | 5 | 497 | |||
| NM_029528.5 | NP_083804.3 | 5 | 557 | |||
| GenBank mRNA | AK018206.1 | - | 5 | 496 | ||
| AK018325.1 | - | 5 | 503 | |||
| AK047763.1 | - | 5 | 483 | |||
| AK158869.1 | - | 5 | 450 | |||
| AK163654.1 | - | 4 | 442 | |||
| BC115595.1 | - | 4 | 287 | |||
| BC115596.1 | - | 4 | 287 | |||

Fam131b
Genomic Map