siRNA Details
| Gene Symbol: Fam131b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113327.2 | NP_001106798.1 | 4 | 590 | ||
| NM_001286584.1 | NP_001273513.1 | 5 | 566 | |||
| NM_029528.5 | NP_083804.3 | 5 | 626 | |||
| GenBank mRNA | AK018206.1 | - | 5 | 565 | ||
| AK018325.1 | - | 5 | 572 | |||
| AK047763.1 | - | 5 | 552 | |||
| AK158869.1 | - | 5 | 519 | |||
| AK163654.1 | - | 4 | 511 | |||
| BC115595.1 | - | 4 | 356 | |||
| BC115596.1 | - | 4 | 356 | |||

Fam131b
Genomic Map