siRNA Details
| Gene Symbol: Cyp2d26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029562.2 | NP_083838.1 | 5 | 818 | ||
| GenBank mRNA | AK004915.1 | - | 5 | 819 | ||
| BC023241.1 | - | 5 | 807 | 4222538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2d26| Gene Symbol: Cyp2d26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029562.2 | NP_083838.1 | 5 | 818 | ||
| GenBank mRNA | AK004915.1 | - | 5 | 819 | ||
| BC023241.1 | - | 5 | 807 | 4222538 | ||