siRNA Details
| Gene Symbol: Lyrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285959.1 | NP_001272888.1 | 4 | 511 | ||
| NM_001285960.1 | NP_001272889.1 | 4 | 644 | |||
| NM_029610.3 | NP_083886.1 | 4 | 895 | |||
| GenBank mRNA | AK009135.1 | - | 4 | 898 | ||
| BC011339.1 | - | 4 | 685 | 3978191 | ||
| DV043140.1 | - | 3 | 669 | |||

Lyrm1
Genomic Map