siRNA Details
| Gene Symbol: Ier5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030244.3 | NP_084520.3 | 1 | 999 | ||
| GenBank mRNA | AK012145.1 | - | 1 | 1013 | ||
| AK034055.1 | - | 1022 | ||||
| BC003464.1 | - | 1 | 998 | 2648900 | ||
| BC010234.1 | - | 1 | 999 | 3499467 | ||
| BC040084.1 | - | 1 | 999 | 3499467 | ||

Ier5l
Genomic Map