siRNA Details
| Gene Symbol: SRPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170750.1 | NP_001164221.1 | 8 | 1246 | ||
| NM_001170751.1 | NP_001164222.1 | 8 | 1129 | |||
| NM_006307.4 | NP_006298.1 | 9 | 1306 | |||
| XM_017029893.1 | XP_016885382.1 | 9 | 1245 | |||
| GenBank mRNA | AK125542.1 | - | 8 | 1227 | ||
| AK289430.1 | - | 9 | 1253 | |||
| AK298802.1 | - | 8 | 1065 | |||
| BC020684.1 | - | 9 | 1261 | 4731946 | ||
| EU668359.1 | - | 9 | 1206 | |||
| U61374.1 | - | 9 | 1206 | |||
| U78093.1 | - | 9 | 1213 | |||

SRPX
Genomic Map