siRNA Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031373.3 | NP_113550.3 | 7 | 672 | ||
| GenBank mRNA | AF303894.1 | - | 7 | 620 | ||
| AK008230.1 | - | 7 | 637 | |||
| AK149146.1 | - | 7 | 619 | |||
| AK150115.1 | - | 7 | 604 | |||
| BC003875.1 | - | 7 | 603 | 3495186 | ||
| BC049965.1 | - | 7 | 618 | 3155091 | ||

Ogfr
Genomic Map