siRNA Details
| Gene Symbol: Lpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080420.2 | NP_536345.2 | 8 | 1321 | ||
| GenBank mRNA | BC016212.1 | - | 8 | 1328 | 4924899 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpo| Gene Symbol: Lpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080420.2 | NP_536345.2 | 8 | 1321 | ||
| GenBank mRNA | BC016212.1 | - | 8 | 1328 | 4924899 | |