siRNA Details
| Gene Symbol: Msl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163833.1 | NP_001157305.1 | 2 | 1813 | ||
| GenBank mRNA | AK006842.1 | - | 1 | 1214 | ||
| AK044179.1 | - | 1 | 1213 | |||
| AK155382.1 | - | 2 | 1799 | |||
| AK172141.1 | - | 195 | ||||
| BC013191.1 | - | 1 | 1415 | |||
| BC037487.1 | - | 2 | 1824 | |||

Msl3l2
Genomic Map