siRNA Details
| Gene Symbol: Errfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133753.1 | NP_598514.1 | 3 | 411 | ||
| GenBank mRNA | AK004851.1 | - | 3 | 430 | ||
| AK139780.1 | - | 3 | 427 | |||
| AK166791.1 | - | 3 | 430 | |||
| AK167156.1 | - | 3 | 370 | |||
| AK167359.1 | - | 3 | 430 | |||
| AK205245.1 | - | 2 | 242 | |||
| AK212670.1 | - | 2 | 262 | |||
| BC005546.1 | - | 3 | 411 | 3585640 | ||
| BC057646.1 | - | 3 | 429 | 4194124 | ||

Errfi1
Genomic Map